Alfa Aesar™ Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length, 99% (metals basis), Si nominal 1.5%

High resistivity material for electronics industry

Manufacturer: Alfa Aesar™ 4043706

Quantity 5g

 View more versions of this product

Catalog No. AA4043706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11106-97-1
    MDL Number MFCD00673110
    Chemical Name or Material Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00673110
    CAS 11106-97-1
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    TSCA TSCA
    Physical Form Pieces
    Health Hazard 1 Warning
    Weight ≈0.21g/piece
    Concentration Si nomin.al 1.5%
    Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
    Quantity 5g
    Chemical Name or Material Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length
    Assay 99%, (metals basis)

    GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit