Alfa Aesar™ Nickel Chromium powder, -140+325 mesh, 97% (metals basis)

Manufacturer: Alfa Aesar™ 1310836

Quantity 500g

 View more versions of this product

Catalog No. AA1310836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11106-97-1
    MDL Number MFCD00673110
    UN Number UN3178
    Synonym Nichrome™
    Chemical Name or Material Nickel Chromium powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    UN Number UN3178
    Mesh Size −140+325 Mesh
    Health Hazard 2 GHS H Statement
    H228-H351-H317
    Flammable solid.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Density 3.1g/mL
    CAS 11106-97-1
    Synonym Nichrome™
    Odor Odorless
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD00673110
    Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
    Quantity 500g
    Chemical Name or Material Nickel Chromium powder
    Assay 97%, (metals basis)

    GHS H Statement
    H228-H351-H317
    Flammable solid.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit