Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel Chromium powder, -140+325 mesh, 97% (metals basis)

Manufacturer:  Alfa Aesar™ 1310822

Quantity 100g

 View more versions of this product

Catalog No. AA1310822

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

11106-97-1
UN3178
Nickel Chromium powder
MFCD00673110
Nichrome?

Specifications

Ni:Cr; 80:20 wt%
11106-97-1
Powder
UN3178
Nichrome?
Nickel Chromium powder
100g
3.1g/mL
MFCD00673110
−140+325 Mesh
Odorless
97%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H351-H317
Flammable solid.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit