Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 4236609

Quantity 10g

 View more versions of this product

Catalog No. AA4236609


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

11106-97-1
MFCD00673110
Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length

Specifications

Ni:Cr; 80:20 wt%
10g
Slug
Odorless
99.95%, (metals basis)
≈1.7g/slug
11106-97-1
MFCD00673110
Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length

Safety and Handling

SDS & Safety Documents

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit