Learn More
Humboldt Nickel-Chromium Triangles
Manufacturer:  Humboldt H23970
 View more versions of this product
Catalog No. 15-260C
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use with platinum crucibles. Resist oxidation and will not contaminate platinum. 80-20 nickel-chromium alloy, contains no iron.