Learn More
Nickel Chromium wire, 0.41mm (0.016in) dia, Thermo Scientific Chemicals
Description
These alloys are typically used in resistance wire. They are also used in some dental restorations (fillings). It is commonly used in ceramic as an internal support. Nichrome wire is used for its ability to withstand the high temperatures that occur when clay work is fired in a kiln.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cr; 80:20 wt% |
Quantity | 25 m |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Weight | ≈1.09g/m |
Form | Wire |
Chemical Name or Material | Nickel Chromium wire, 0.41mm (0.016 in.) dia. |
Safety and Handling
Hazard Category | H317-H351 |
Hazard Statement | GHS H Statement H351-H317 Suspected of causing cancer. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.