Nickel-Chromium Wire

For making inoculating needles and loops

Manufacturer: ARCOR ELECTRONIC COMPANY 22S

 View more versions of this product

Catalog No. 13-095A

  Check Availability
Add to Cart

    Specifications

    Diameter (English) Outer 0.025 in.
    Diameter (Metric) Outer 0.64mm
    For Use With (Application) For making inoculating needles and loops
    Gauge 22
    Quantity 12 Pk.
    Temperature (English) 2150°F
    Temperature (Metric) 1177°C
    Material Nickel Chromium
    Product Type Nickel Chromium Wire

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit