Learn More
Nickel Copper foil, alloy 400, 0.51mm (0.02in) thick, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045148GT
Description
Nickel Copper foil, alloy 400 finds application in marine engineering, chemical and hydro-carbon processings, heat exchangers, valves and pumps. Large use of it is made in the alkylation units, namely in the reacting section in contact with concentrated hydrofluoric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Copper | |
Foil | |
MFCD00801098 | |
Insoluble in water. |
Alloy 400 | |
Ni:Cu; 67:33 wt.% | |
150 x 150 mm | |
Foil |
Safety and Handling
Wear suitable protective clothing and gloves.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only