Learn More
Nickel Copper foil, alloy 400, 0.81mm (0.032 in.) thick, Thermo Scientific Chemicals
$126.00 - $344.65
Specifications
Chemical Name or Material | Nickel Copper foil, Alloy 400 |
---|---|
Name Note | 0.81mm (0.032 in.) thick |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cu; 67:33 wt% |
MDL Number | MFCD00801098 |
Form | Foil |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA45169GT
|
Thermo Scientific Chemicals
045169GT |
150 x 150 mm |
Each for $126.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AA45169HB
|
Thermo Scientific Chemicals
45169HB |
300 x 300 mm |
Each
for $344.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Copper foil, Alloy 400 | |
Ni:Cu; 67:33 wt% | |
Foil |
0.81mm (0.032 in.) thick | |
MFCD00801098 |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.