Alfa Aesar™ Nickel Copper foil, alloy 400, 0.81mm (0.032 in.) thick

Manufacturer: Alfa Aesar™ 45169GT

Quantity 150x150mm

 View more versions of this product

Catalog No. AA45169GT

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00801098
    Chemical Name or Material Nickel Copper foil, Alloy 400, 0.81mm (0.032 in.) thick

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Foil
    Health Hazard 2 GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Avg. Mol. Wt. or Mol. Wt. Range Ni:Cu; 67:33 wt%
    Quantity 150x150mm
    TSCA TSCA
    MDL Number MFCD00801098
    Health Hazard 1 Warning
    Chemical Name or Material Nickel Copper foil, Alloy 400, 0.81mm (0.032 in.) thick

    GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit