Alfa Aesar™ Nickel dimethylglyoxime, 99%

Paint pigment

Manufacturer: Alfa Aesar™ 3945818

Quantity 50g

 View more versions of this product

Catalog No. AA3945818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13478-93-8
    Formula Weight 288.94
    MDL Number MFCD00136028
    Chemical Name or Material Nickel dimethylglyoxime

    Specifications

    Formula Weight 288.94
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Melting Point 250°C (subl.)
    Health Hazard 1 Warning
    MDL Number MFCD00136028
    Quantity 50g
    Solubility Information Insoluble in water; Soluble in mineral acids
    CAS 13478-93-8
    Health Hazard 2 GHS H Statement
    H351-H315-H319-H317-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Chemical Name or Material Nickel dimethylglyoxime
    Assay 99%

    GHS H Statement
    H351-H315-H319-H317-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit