Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel dimethylglyoxime, 99%

Click to view available options
Quantity:
2g
10g
50g
13478-93-8
C8H16N4NiO4
290.933
MFCD00136028
CNOULSXJEKYGTK-FDGPNNRMSA-N
nickel dimethylglyoxime,,nickel; n-z-3-nitrosobut-2-en-2-yl hydroxylamine
15639912
nickel;N-[(Z)-3-nitrosobut-2-en-2-yl]hydroxylamine
CC(=C(C)N=O)NO.CC(=C(C)N=O)NO.[Ni]
13478-93-8
C8H16N4NiO4
290.933
MFCD00136028
CNOULSXJEKYGTK-FDGPNNRMSA-N
nickel dimethylglyoxime,,nickel; n-z-3-nitrosobut-2-en-2-yl hydroxylamine
15639912
nickel;N-[(Z)-3-nitrosobut-2-en-2-yl]hydroxylamine
CC(=C(C)N=O)NO.CC(=C(C)N=O)NO.[Ni]

Paint pigment

Quantity 2g
Melting Point 250°C (sublimation)
Solubility Information Insoluble in water; Soluble in mineral acids
Formula Weight 288.94
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Nickel dimethylglyoxime

GHS H Statement
H351-H315-H319-H317-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c

H315-H317-H319-H335-H350

EINECSNumber : 236-782-7

TSCA : No

Recommended Storage : Ambient temperatures