Alfa Aesar™ Nickel etchant for electrodeposited nickel films

High purity etchant for electrodeposited nickel films

Manufacturer: Alfa Aesar™ 44580LV

Quantity 1 gal.

 View more versions of this product

Catalog No. AA44580LV

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD09039236
    UN Number UN1760
    Chemical Name or Material Nickel etchant for electrodeposited nickel films

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD09039236
    Health Hazard 2 GHS H Statement
    H314-H351-H317-H319
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Quantity 1 gal.
    UN Number UN1760
    Chemical Name or Material Nickel etchant for electrodeposited nickel films

    GHS H Statement
    H314-H351-H317-H319
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Causes serious eye irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit