Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel foil, 0.025mm (0.001in) thick, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number AA10254FI
Click to view available options
Quantity:
50 x 50 mm
50 x 250 mm
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 x 50 mm
Percent Purity 99.99%
Odor Odorless
Weight ≈0.56g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Nickel foil, 0.025mm (0.001 in.) thick
Hazard Category H317-H351-H372
Hazard Statement GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit