Nickel Foil, 0.5mm (0.02 in.) thick, 99.5% (Metals basis), Alfa Aesar™

Form: Approx.11.1g/50 x 50mm

Manufacturer: Alfa Aesar 44823GH

Quantity 100 x 100mm

 View more versions of this product

Catalog No. AA44823GH

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-02-0
    Physical Form Foil, 0.5mm (0.02 in.) thick
    Assay Percent Range 99.5% (Metals basis)
    MDL Number MFCD00011137
    Chemical Name or Material Nickel

    Specifications

    Quantity 100 x 100mm
    Color Silver-gray
    EINECS Number 231-111-4
    Physical Form Foil, 0.5mm (0.02 in.) thick
    Assay Percent Range 99.5% (Metals basis)
    MDL Number MFCD00011137
    CAS 7440-02-0
    Odor Odorless
    Health Hazard 1 H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
    Appearance Silver gray foil
    Chemical Name or Material Nickel

    H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit