Nickel gauze, 100 mesh woven, Alfa Aesar™

Form: Wire Cloth; Open area: 36%; Width of opening: 0.006 in.

Manufacturer: Alfa Aesar 44128HB

Quantity 300 x 300mm

 View more versions of this product

Catalog No. AA44128HB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-02-0
    Physical Form Gauze
    MDL Number MFCD00011137
    Chemical Name or Material Nickel

    Specifications

    Quantity 300 x 300mm
    Color Silver-gray
    EINECS Number 231-111-4
    Physical Form Gauze
    MDL Number MFCD00011137
    CAS 7440-02-0
    Odor Odorless
    Health Hazard 1 H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
    Appearance Silver gray guaze
    Chemical Name or Material Nickel

    H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit