Alfa Aesar™ Nickel hydroxyacetate, Ni 27-29%

Manufacturer: Alfa Aesar™ 3945622

Quantity 100g

 View more versions of this product

Catalog No. AA3945622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41587-84-2
    Linear Formula Ni(OOCCH2OH)2
    Formula Weight 208.78
    MDL Number MFCD00070414
    UN Number UN3077
    Chemical Name or Material Nickel hydroxyacetate

    Specifications

    Formula Weight 208.78
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    MDL Number MFCD00070414
    UN Number UN3077
    Linear Formula Ni(OOCCH2OH)2
    CAS 41587-84-2
    Health Hazard 1 Danger
    Concentration Ni 27 to 29%
    Quantity 100g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Color Green
    Chemical Name or Material Nickel hydroxyacetate

    GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit