Alfa Aesar™ Nickel(II) 1,1,1-trifluoro-2,4-pentanedionate dihydrate

Manufacturer: Alfa Aesar™ 3944903

Quantity 1g

 View more versions of this product

Catalog No. AA3944903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 55534-89-9
    Formula Weight 400.90 (364.87 Anhydrous)
    MDL Number MFCD00150656
    Synonym Nickel(II) 1,1,1-trifluoroacetylacetonate dihydrate
    Chemical Name or Material Nickel(II) 1,1,1-trifluoro-2,4-pentanedionate dihydrate

    Specifications

    Formula Weight 400.90 (364.87 Anhydrous)
    Health Hazard 2 GHS H Statement
    H350-H302-H315-H319-H317
    May cause cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Physical Form Powder
    CAS 55534-89-9
    Quantity 1g
    Health Hazard 1 Danger
    MDL Number MFCD00150656
    Color Green
    Synonym Nickel(II) 1,1,1-trifluoroacetylacetonate dihydrate
    Chemical Name or Material Nickel(II) 1,1,1-trifluoro-2,4-pentanedionate dihydrate

    GHS H Statement
    H350-H302-H315-H319-H317
    May cause cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit