Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel(II) 2,4-pentanedionate, 95%

Catalyst and U.V. stabilizer

Manufacturer:  Alfa Aesar™ 1881114

Quantity 25g

 View more versions of this product

Catalog No. AA1881114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3264-82-2
MFCD00000024
14,6500
Nickel(II) 2,4-pentanedionate
256.91
4157970
Bis(acetylacetonato)nickel(II); Nickel(II) acetylacetonate

Specifications

3264-82-2
Powder
≈230°C (dec.)
4157970
25g
Odorless
Soluble in water, alcohol, toluene
95%
Hygroscopic
256.91
MFCD00000024
Green
14,6500
Bis(acetylacetonato)nickel(II); Nickel(II) acetylacetonate
Nickel(II) 2,4-pentanedionate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

Danger

RTECSNumber : SA2100000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit