Alfa Aesar™ Nickel(II) 2,4-pentanedionate, 95%

Catalyst and U.V. stabilizer

Manufacturer: Alfa Aesar™ 1881106

Quantity 5g

 View more versions of this product

Catalog No. AA1881106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3264-82-2
    Formula Weight 256.91
    MDL Number MFCD00000024
    Beilstein 4157970
    Merck Index 14,6500
    Synonym Bis(acetylacetonato)nickel(II); Nickel(II) acetylacetonate
    Chemical Name or Material Nickel(II) 2,4-pentanedionate

    Specifications

    Formula Weight 256.91
    Beilstein 4157970
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H350-H302-H317
    May cause cancer.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Sensitivity Hygroscopic
    Physical Form Powder
    MDL Number MFCD00000024
    CAS 3264-82-2
    Solubility Information Soluble in water, alcohol, toluene
    Health Hazard 1 Danger
    TSCA TSCA
    Melting Point ≈230°C (dec.)
    Color Green
    Merck Index 14,6500
    Quantity 5g
    Synonym Bis(acetylacetonato)nickel(II); Nickel(II) acetylacetonate
    RTECS Number SA2100000
    Chemical Name or Material Nickel(II) 2,4-pentanedionate
    Assay 95%

    GHS H Statement
    H350-H302-H317
    May cause cancer.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit