Alfa Aesar™ Nickel(II) 2-ethylhexanoate monoisopropoxide, 99+% (m.b.), 5% w/v in IPA, pkgd under Ar in rslble ChemSeal bottles

Manufacturer: Alfa Aesar™ 42380AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42380AE

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 261
    MDL Number MFCD01863473
    UN Number UN1219
    Synonym Nickel(II) 2-ethylhexano-isopropoxide
    Chemical Name or Material Nickel (II) 2-ethylhexanoate monoisopropoxide

    Specifications

    Formula Weight 261
    Health Hazard 2 GHS H Statement
    H225-H351-H361-H315-H319-H317-H336
    Highly flammable liquid and vapor.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P280h-P243-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/clothing.
    Take precautionary measures against static discharge.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Packaging ChemSeal™ bottles
    Physical Form Liquid
    MDL Number MFCD01863473
    Quantity 100mL
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Sensitivity Moisture Sensitive
    Concentration 5% w/v in isopropanol
    UN Number UN1219
    Synonym Nickel(II) 2-ethylhexano-isopropoxide
    Storage Note 1
    • Packaged under Argon
    Chemical Name or Material Nickel (II) 2-ethylhexanoate monoisopropoxide
    Assay ≥99% (m.b.)

    GHS H Statement
    H225-H351-H361-H315-H319-H317-H336
    Highly flammable liquid and vapor.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280h-P243-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/clothing.
    Take precautionary measures against static discharge.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit