Learn More
Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
It is used in electroplating, anodize coatings; textile dyeing mordant; Catalyst. Also, used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Green |
Physical Form | Crystalline Aggregates |
Quantity | 5 g |
Linear Formula | Ni(CH3COO)2·4H2O |
UN Number | UN3077 |
Beilstein | 3730764 |
Merck Index | 14,6499 |
Solubility Information | Soluble in water and methanol. Insoluble in diethyl ether,n-octanol. |
Formula Weight | 248.84 (176.79 Anhydrous) |
Show More |
Safety and Handling
Hazard Category | H302+H332-H317-H334-H335-H341-H350i-H360D-H372 |
Hazard Statement | GHS H Statement H334-H350-H360-H372-H341-H302-H332-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 206-761-7 |
RTECSNumber | QR6126000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.