Alfa Aesar™ Nickel(II) bromide, anhydrous, 99%

Manufacturer: Alfa Aesar™ 5311518

Quantity 50g

 View more versions of this product

Catalog No. AA5311518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13462-88-9
    Molecular Formula Br2Ni
    Linear Formula NiBr2
    Formula Weight 218.53
    MDL Number MFCD00011141
    UN Number UN3077
    Merck Index 14,6502
    Chemical Name or Material Nickel (II) bromide, Anhydrous

    Specifications

    Formula Weight 218.53
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P273-P201-P309-P310-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula NiBr2
    Density 5.098g/mL at 27°C
    Odor Odorless
    Physical Form Crystalline
    Sensitivity Hygroscopic
    Quantity 50g
    TSCA TSCA
    Molecular Formula Br2Ni
    UN Number UN3077
    Merck Index 14,6502
    Health Hazard 1 Danger
    MDL Number MFCD00011141
    CAS 13462-88-9
    Melting Point 963°C
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Nickel (II) bromide, Anhydrous
    Assay 99%

    GHS H Statement
    H334-H350-H360-H372-H341-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.

    GHS P Statement
    P273-P201-P309-P310-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit