Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) bromide hydrate, 98%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
207569-11-7,13462-88-9
Br2Ni·xH2O
218.53
MFCD00149806
LQJMXNQEJAVYNB-UHFFFAOYSA-L
nickel ii bromide hydrate,dibromonickel hydrate,nickel bromide hydrate,br2ni.h2o,ksc566q3p,nickel ii bromide monohydrate,nickel bromide nibr2 ,monohydrate 9ci
57377118
dibromonickel;hydrate
O.[Ni](Br)Br
207569-11-7,13462-88-9
Br2Ni·xH2O
218.53
MFCD00149806
LQJMXNQEJAVYNB-UHFFFAOYSA-L
nickel ii bromide hydrate,dibromonickel hydrate,nickel bromide hydrate,br2ni.h2o,ksc566q3p,nickel ii bromide monohydrate,nickel bromide nibr2 ,monohydrate 9ci
57377118
dibromonickel;hydrate
O.[Ni](Br)Br
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Linear Formula NiBr2·xH2O
Fieser 04,351
Merck Index 10, 6340
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ammonium hydroxide,soluble in alcohol and ether
Formula Weight 218.53
Physical Form Crystalline Powder or Needles
Percent Purity 98%
Grade Analysis
Cadmium (Cd) 5ppm max.
Calcium (Ca) 50ppm max.
Cobalt (Co) 100ppm max.
Copper (Cu) 10ppm max.
Iron (Fe) 20ppm max.
Magnesium (Mg) 20ppm max.
Packaging Glass bottle
Potassium (K) 20ppm max.
Sodium (Na) 500ppm max.
Solubility Solubility in water: soluble in water. Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether
Sulfate 50ppm max.
Water 21% max. (K.F.)
Color Orange
Melting Point 963.0°C
pH 5 to 7 (5% aq. soln.)
Quantity 500g
Chemical Name or Material Nickel(II) bromide hydrate, 98%
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
M

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain speci

GHS Signal Word: Danger