Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) chloride, 98%, Acros Organics

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
7718-54-9
Cl2Ni
129.6
MFCD00011142
QMMRZOWCJAIUJA-UHFFFAOYSA-L
24385
CHEBI:34887
dichloronickel
Cl[Ni]Cl
7718-54-9
Cl2Ni
129.6
MFCD00011142
QMMRZOWCJAIUJA-UHFFFAOYSA-L
24385
CHEBI:34887
dichloronickel
Cl[Ni]Cl
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula NiCl2
Merck Index 15, 6590
Solubility Information Solubility in water: slightly soluble
Formula Weight 129.6
Physical Form Powder
Percent Purity 98%
Packaging Glass bottle
Solubility Solubility in water: slightly soluble.
Color Brown to Orange
Melting Point 1001.0°C
Quantity 250g
Chemical Name or Material Nickel(II) chloride
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger

EINECSNumber : 231-743-