Nickel(II) chloride, anhydrous, 99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1468722

Quantity 100g

 View more versions of this product

Catalog No. AA1468722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7718-54-9
    Physical Form Powder
    Assay Percent Range 99% (metals basis)
    Molecular Formula Cl2Ni
    Formula Weight 129.6
    MDL Number MFCD00011142
    Chemical Name or Material Nickel(II) chloride, anhydrous

    Specifications

    Physical Form Powder
    Formula Weight 129.6
    Quantity 100g
    Color Yellow to orange
    DOT Information Hazard Class: 6.1, Packing Group: III
    Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
    Density 3.55g/cm3
    EINECS Number 231-743-0
    Anhydrous or Hydrate Anhydrous
    Assay Percent Range 99% (metals basis)
    Molecular Formula Cl2Ni
    Appearance Yellow to orange powder
    MDL Number MFCD00011142
    CAS 7718-54-9
    Vapor Pressure 1.33 hPa (1mm Hg) at 20°C
    Health Hazard 1 H301-H331-H315-H317-H334-H350-H360-H372-H341-H400-H410: Toxic if swallowed. Toxic if inhaled. Causes skin irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Nickel(II) chloride, anhydrous

    Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H301-H331-H315-H317-H334-H350-H360-H372-H341-H400-H410: Toxic if swallowed. Toxic if inhaled. Causes skin irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit