Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) chloride hexahydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2kg
Packaging:
Glass bottle
7791-20-0,7718-54-9
Cl2Ni·6H2O
237.71
MFCD00149809
7791-20-0,7718-54-9
Cl2Ni·6H2O
237.71
MFCD00149809
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Complexometry)
Linear Formula NiCl2·6H2O
Fieser 14,351; 15,96; 17,86
Merck Index 15, 6590
Solubility Information Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
Formula Weight 237.71
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Cadmium (Cd) 10ppm max.
Calcium (Ca) 50ppm max.
Cobalt (Co) 60ppm max.
Copper (Cu) 10ppm max.
Iron (Fe) 20ppm max.
Lead (Pb) 10ppm max.
Magnesium (Mg) 10ppm max.
Packaging Glass bottle
Sodium (Na) 50ppm max.
Solubility Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
Sulfate 50ppm max.
Zinc (Zn) 10ppm max.
Color Green
pH 4 to 6 (5% soln. at 20°C)
Quantity 2kg
Chemical Name or Material Nickel(II) chloride hexahydrate, 97%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid relea

GHS Signal Word: Danger

RTECSNumber : QR6480000