Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel(II) chloride hexahydrate, 97%, ACROS Organics™
Click to view available options
Quantity:
25g
500g
2kg
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Assay Percent Range | 96% min. (Complexometry) |
Linear Formula | NiCl2·6H2O |
Fieser | 14,351; 15,96; 17,86 |
Merck Index | 15, 6590 |
Solubility Information | Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol |
Formula Weight | 237.71 |
Physical Form | Crystals or Crystalline Powder |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid relea
GHS Signal Word: Danger
RTECSNumber : QR6480000