Learn More
Nickel(II) chloride hexahydrate, 99.3% (metals basis), Thermo Scientific Chemicals
Description
It is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is used to prepare a wide array of complexes since water ligands can be easily replaced by amines, thioether, ammonia, and thiolates. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 140°C-H2O |
Physical Form | Crystalline |
Quantity | 2 kg |
Assay Percent Range | 99.3% (metals basis) |
Solubility Information | Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide. |
Formula Weight | 237.71 (129.60 Anhydrous) |
Odor | Odorless |
Appearance | Green crystals |
Density | 1.92 |
Chemical Name or Material | Nickel(II) chloride hexahydrate |
Safety and Handling
Hazard Category | H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372 |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c |
DOTInformation | DOT Class: 6.1, Packaging Group: III |
EINECSNumber | 231-743-0 |
RTECSNumber | QR6480000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.