Nickel(II) chloride hexahydrate, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 5313130

Quantity 250g

 View more versions of this product

Catalog No. AA5313130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7791-20-0
    Physical Form Crystalline
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula Cl2Ni·6H2O
    Formula Weight 237.71
    MDL Number MFCD00149809
    Chemical Name or Material Nickel(II) chloride hexahydrate

    Specifications

    Physical Form Crystalline
    Formula Weight 237.71
    Quantity 250g
    Color Green
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 231-743-0
    Odor Odorless
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula Cl2Ni·6H2O
    MDL Number MFCD00149809
    CAS 7791-20-0
    Health Hazard 1 H301-H331-H315-H317-H334-H350-H360-H372-H341-H400-H410: Toxic if swallowed. Toxic if inhaled. Causes skin irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Nickel(II) chloride hexahydrate

    H301-H331-H315-H317-H334-H350-H360-H372-H341-H400-H410: Toxic if swallowed. Toxic if inhaled. Causes skin irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit