Alfa Aesar™ Nickel(II) fluoride tetrahydrate, 98+%

Manufacturer: Alfa Aesar™ 1413230

Quantity 250g

 View more versions of this product

Catalog No. AA1413230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13940-83-5
    Formula Weight 168.77 (96.71 Anhydrous)
    MDL Number MFCD00016263
    UN Number UN3288
    Chemical Name or Material Nickel(II) fluoride tetrahydrate

    Specifications

    Formula Weight 168.77 (96.71 Anhydrous)
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00016263
    UN Number UN3288
    Quantity 250g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Odor Odorless
    CAS 13940-83-5
    RTECS Number QR6825000
    Health Hazard 1 Danger
    Chemical Name or Material Nickel(II) fluoride tetrahydrate
    Assay ≥98%

    GHS H Statement
    H334-H350-H360-H372-H341-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit