Nickel(II) formate dihydrate, Alfa Aesar™

For Nickel Catalyst Preparations

Manufacturer: Alfa Aesar 1380122

Quantity 100g

 View more versions of this product

Catalog No. AA1380122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15694-70-9
    Physical Form Powder
    Molecular Formula Ni(HCO2)2·2H2O
    Formula Weight 184.77 (148.74 Anhydrous)
    MDL Number MFCD00153034
    Chemical Name or Material Nickel(II) formate dihydrate

    Specifications

    Physical Form Powder
    Formula Weight 184.77 (148.74 Anhydrous)
    Quantity 100g
    Color Green
    DOT Information Hazard Class: 9, Packing Group: III
    Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
    Decomposition Information In the range 180° to 200°C decomposes to Ni,CO,CO2
    Density 2.15g/cm3
    EINECS Number 222-101-0
    Anhydrous or Hydrate Dihydrate
    Molecular Formula Ni(HCO2)2·2H2O
    Appearance Green powder
    MDL Number MFCD00153034
    CAS 15694-70-9
    Health Hazard 1 H334-H350-H360-H372-H317-H341-H400-H410: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Nickel(II) formate dihydrate

    Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H334-H350-H360-H372-H317-H341-H400-H410: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit