Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel(II) hydroxide, Ni 61%

Manufacturer:  Alfa Aesar™ 12517A7

Quantity 5kg

 View more versions of this product

Catalog No. AA12517A7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12054-48-7
92.7
UN3077
Ni(OH)2
MFCD00011140
Nickel(II) hydroxide

Specifications

12054-48-7
Ni(OH)2
MFCD00011140
Ni 61%
Odorless
230°C (dec.)
Nickel(II) hydroxide
Powder
92.7
UN3077
4.1g/mL
Green
5kg

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P261-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : QR7040000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit