Alfa Aesar™ Nickel(II) hydroxide, Ni 61%

Manufacturer: Alfa Aesar™ 1251706

Quantity 5g

 View more versions of this product

Catalog No. AA1251706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12054-48-7
    Linear Formula Ni(OH)2
    Formula Weight 92.7
    MDL Number MFCD00011140
    UN Number UN3077
    Chemical Name or Material Nickel(II) hydroxide

    Specifications

    Formula Weight 92.7
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H315-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 12054-48-7
    UN Number UN3077
    Color Green
    Melting Point 230°C (dec.)
    RTECS Number QR7040000
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00011140
    Health Hazard 1 Danger
    Density 4.1g/mL
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Physical Form Powder
    Quantity 5g
    Linear Formula Ni(OH)2
    Concentration Ni 61%
    Chemical Name or Material Nickel(II) hydroxide

    GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H315-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit