Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel(II) lactate tetrahydrate, 98%

Click to view available options
Quantity:
25g
100g
500g
C6H10NiO6·4H2O
MFCD02683106
C6H10NiO6·4H2O
MFCD02683106
UN Number UN3077
Formula Weight 308.91 (236.85 Anhydrous)
Percent Purity 98%
Quantity 100g
Chemical Name or Material Nickel(II) lactate tetrahydrate

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.