Alfa Aesar™ Nickel(II) octanoate, in mineral spirits (8% Ni)

Manufacturer: Alfa Aesar™ 3947922

Quantity 100g

 View more versions of this product

Catalog No. AA3947922

  Check Availability
Add to Cart

    Chemical Identifiers

    Linear Formula [CH3(CH2)6COO]2Ni
    Formula Weight 345.1
    MDL Number MFCD00064239
    UN Number UN1993
    Synonym Octanoic acid nickel(II) salt
    Chemical Name or Material Nickel(II) octanoate, In mineral spirits

    Specifications

    Formula Weight 345.1
    Health Hazard 2 GHS H Statement
    H334-H340-H350-H360-H372-H304-H226-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula [CH3(CH2)6COO]2Ni
    UN Number UN1993
    Quantity 100g
    Physical Form Liquid
    Density 0.95g/mL
    Concentration 8% Ni
    Color Green
    Flash Point 40° (104°F)
    Synonym Octanoic acid nickel(II) salt
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    TSCA TSCA
    MDL Number MFCD00064239
    Health Hazard 1 Danger
    Chemical Name or Material Nickel(II) octanoate, In mineral spirits

    GHS H Statement
    H334-H340-H350-H360-H372-H304-H226-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit