Alfa Aesar™ Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis)

Manufacturer: Alfa Aesar™ 4075714

Quantity 25g

 View more versions of this product

Catalog No. AA4075714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 126956-48-7
    Linear Formula NiC2O4·xH2O (x≈2)
    Formula Weight 146.73 (Anhydrous)
    MDL Number MFCD00167155
    UN Number UN3077
    Merck Index 14,6514
    Chemical Name or Material Nickel(II) oxalate hydrate
    Grade Puratronic™

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 146.73 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00167155
    UN Number UN3077
    Merck Index 14,6514
    Color Green
    Quantity 25g
    Health Hazard 1 Danger
    TSCA TSCA
    Linear Formula NiC2O4·xH2O (x≈2)
    CAS 126956-48-7
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Health Hazard 2 GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Chemical Name or Material Nickel(II) oxalate hydrate
    Assay 99.9985%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit