Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051212
Click to view available options
Quantity:
5 g
25 g
126956-48-7
C2NiO4
146.71
MFCD00167155
DOLZKNFSRCEOFV-UHFFFAOYSA-L
nickel ii oxalate hydrate, acmc-1c3o8, nickel 2+ hydrate oxalate, nickel ii oxalate hydrate, puratronic, nickel 2+ ion hydrate oxalate, nickel 2+ ethanedioate-water 1/1/1, nickel, ethanedioato 2--ko1,ko2-, hydrate 9ci
16213068
[Ni++].[O-]C(=O)C([O-])=O
126956-48-7
C2NiO4
146.71
MFCD00167155
DOLZKNFSRCEOFV-UHFFFAOYSA-L
nickel ii oxalate hydrate, acmc-1c3o8, nickel 2+ hydrate oxalate, nickel ii oxalate hydrate, puratronic, nickel 2+ ion hydrate oxalate, nickel 2+ ethanedioate-water 1/1/1, nickel, ethanedioato 2--ko1,ko2-, hydrate 9ci
16213068
[Ni++].[O-]C(=O)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Green
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula NiC2O4·xH2O (x≈2)
UN Number UN3077
Merck Index 14,6514
IUPAC Name nickel(2+);oxalate;hydrate
Formula Weight 146.73 (Anhydrous)
Percent Purity 99.9985%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Nickel(II) oxalate hydrate
Show More Show Less
Hazard Category H317-H350-H372
Hazard Statement GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 208-933-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit