Nickel(II) oxide, 99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1235918

Quantity 50g

 View more versions of this product

Catalog No. AA1235918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-99-1
    Physical Form -325 Mesh Powder
    Assay Percent Range 99% (metals basis)
    Molecular Formula NiO
    Formula Weight 74.71
    MDL Number MFCD00011145
    Chemical Name or Material Nickel(II) oxide

    Specifications

    Formula Weight 74.71
    Quantity 50g
    EINECS Number 215-215-7
    Physical Form -325 Mesh Powder
    Density 6.67g/mL
    Assay Percent Range 99% (metals basis)
    Molecular Formula NiO
    MDL Number MFCD00011145
    CAS 1313-99-1
    Chemical Name or Material Nickel(II) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit