Alfa Aesar™ Nickel(II) phosphate hydrate

Manufacturer: Alfa Aesar™ 39478-A1

Quantity 1kg

 View more versions of this product

Catalog No. AA39478A1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 27176-17-6
    Formula Weight 366.01 (Anhydrous)
    MDL Number MFCD00049501
    UN Number UN3077
    Merck Index 14,6515
    Chemical Name or Material Nickel(II) phosphate hydrate

    Specifications

    Formula Weight 366.01 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store l
    Physical Form Lump/Powder
    MDL Number MFCD00049501
    CAS 27176-17-6
    UN Number UN3077
    Color Green
    Solubility Information Insoluble in water; Soluble in acids, ammonia
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Merck Index 14,6515
    Quantity 1kg
    Health Hazard 2 GHS H Statement
    H334-H350-H372-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Health Hazard 1 Danger
    Chemical Name or Material Nickel(II) phosphate hydrate

    GHS H Statement
    H334-H350-H372-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store l

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit