Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Nickel(II) phthalocyanine
Click to view available options
Quantity:
5g
25g
Specifications
Color | Blue to Purple |
Melting Point | >300°C |
Quantity | 5g |
Formula Weight | 571.24 |
Physical Form | Powder |
Chemical Name or Material | Nickel(II) phthalocyanine |
Safety and Handling
GHS H Statement
H350-H302-H312-H332
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P261-P272-P280-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H351
EINECSNumber : 237-893-3
TSCA : Yes
Recommended Storage : Ambient temperatures