Alfa Aesar™ Nickel(II) selenide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1310606

Quantity 5g

 View more versions of this product

Catalog No. AA1310606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-05-2
    Formula Weight 137.67
    MDL Number MFCD00049497
    UN Number UN3077
    Chemical Name or Material Nickel(II) selenide

    Specifications

    Formula Weight 137.67
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Insoluble in water; Soluble in aqua regia, HNO3
    Odor Odorless
    UN Number UN3077
    Density 8.46g/mL
    RTECS Number QR8926000
    Physical Form Pieces
    CAS 1314-05-2
    Quantity 5g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Particle Size 6mm & down
    MDL Number MFCD00049497
    Health Hazard 2 GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Chemical Name or Material Nickel(II) selenide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit