Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) sulfate hexahydrate, 99%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
10101-97-0, 7786-81-4
NiO46H2O
262.85
MFCD00149813
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
10101-97-0, 7786-81-4
NiO46H2O
262.85
MFCD00149813
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula NiSO4·6H2O
Merck Index 15,6602
Solubility Information Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol
Formula Weight 262.85
Percent Purity 99%
Grade Analysis
Chloride 50ppm max
Packaging Plastic bottle
Solubility Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol
Melting Point 53.0°C
Quantity 100g
Chemical Name or Material Nickel(II) sulfate hexahydrate, 99%
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
M

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.

GHS Signal Word: Danger

RTECSNumber : QR9600000