Alfa Aesar™ Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)

Manufacturer: Alfa Aesar™ 1082014

Quantity 25g

 View more versions of this product

Catalog No. AA1082014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15244-37-8
    Linear Formula NiSO4·xH2O (x≈6)
    Formula Weight 154.77 (Anhydrous)
    MDL Number MFCD00011146
    UN Number UN3288
    Merck Index 14,6517
    Chemical Name or Material Nickel(II) sulfate hydrate
    Grade Puratronic™

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 154.77 (Anhydrous)
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H315-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P273-P201-P309+P311-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011146
    UN Number UN3288
    Merck Index 14,6517
    pH 4.3 to 4.7
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    TSCA TSCA
    Odor Odorless
    Linear Formula NiSO4·xH2O (x≈6)
    CAS 15244-37-8
    Quantity 25g
    Density 2.070g/mL
    Color Green
    Health Hazard 1 Danger
    Chemical Name or Material Nickel(II) sulfate hydrate
    Assay 99.9985%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H315-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P273-P201-P309+P311-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit