Alfa Aesar™ Nickel(II) tetrafluoroborate hexahydrate

Manufacturer: Alfa Aesar™ 5310822

Quantity 100g

 View more versions of this product

Catalog No. AA5310822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15684-36-3
    Molecular Formula Ni(BF4)2·6H2O
    Formula Weight 340.40 (232.30 Anhydrous)
    MDL Number MFCD00016249
    UN Number UN3077
    Chemical Name or Material Nickel (II) tetrafluoroborate hexahydrate

    Specifications

    Physical Form Lumps
    Formula Weight 340.40 (232.30 Anhydrous)
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Hygroscopic
    Molecular Formula Ni(BF4)2·6H2O
    MDL Number MFCD00016249
    UN Number UN3077
    Health Hazard 1 Danger
    Density 1.5g/mL
    TSCA TSCA
    Odor Odorless
    CAS 15684-36-3
    Quantity 100g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number QR6650000
    Chemical Name or Material Nickel (II) tetrafluoroborate hexahydrate

    GHS H Statement
    H334-H350-H360-H372-H341-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit