Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel(II) tetrafluoroborate hexahydrate, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Blue-Green |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Physical Form | Crystals |
Quantity | 25 g |
Assay Percent Range | 98.5% min. (Complexometry) |
Linear Formula | Ni(BF4)2·6H2O |
Solubility Information | Solubility in water: soluble in water |
IUPAC Name | nickel(2+);ditetrafluoroborate;hexahydrate |
Formula Weight | 340.39 |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | GHS Signal Word: Danger |
missing translation for 'healthHazard2' | GHS H Statement: Causes serious eye damage. Causes severe skin burns and eye damage. May cause cancer. |
missing translation for 'healthHazard3' | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 238-753-4 |
RTECSNumber | QR6650000 |
TSCA | TSCA |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'