Nickel Iron Molybdenum foil, 0.102mm (0.004 in.) thick, annealed

Manufacturer: Alfa Aesar 45092-O

Quantity 300 x 600mm

 View more versions of this product

Catalog No. AA45092O

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Molecular Formula Ni:Fe:Mo
    MDL Number MFCD09910537
    Chemical Name or Material Nickel Iron Molybdenum, 0.102mm (0.004 in.) thick, Annealed

    Specifications

    Quantity 300 x 600mm
    TSCA TSCA
    Physical Form Foil
    Health Hazard 2 GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contam
    Molecular Formula Ni:Fe:Mo
    Odor Odorless
    MDL Number MFCD09910537
    Appearance Silver-colored foil
    Health Hazard 1 H351-H317: Suspected of causing cancer. May cause an allergic skin reaction.
    Chemical Name or Material Nickel Iron Molybdenum, 0.102mm (0.004 in.) thick, Annealed

    GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contam

    H351-H317: Suspected of causing cancer. May cause an allergic skin reaction.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit