Learn More
Nickel, plasma standard solution, Specpure™ Ni 10,000μg/mL, Thermo Scientific Chemicals
Description
Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Concentration | Ni in 5% HNO3 |
Quantity | 100 mL |
UN Number | UN3264 |
Grade | Specpure™ |
Solubility | Miscible with water |
Physical Form | Liquid |
Chemical Name or Material | Nickel plasma standard solution, Ni 10000μg/mL |
Safety and Handling
Hazard Category | H290-H314-H317-H335-H350-H373 |
Hazard Statement | GHS H Statement H314-H351-H373-H317 Causes severe skin burns and eye damage. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P234-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.