Alfa Aesar™ Nickel plating solution, bright finish

Manufacturer: Alfa Aesar™ 42027-K2

Quantity 1L

 View more versions of this product

Catalog No. AA42027K2

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00011137
    UN Number UN3082
    Chemical Name or Material Nickel plating solution, Bright finish

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container to local regulations
    UN Number UN3082
    Odor Odorless
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H315-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prol
    Color Green
    Quantity 1L
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00011137
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Chemical Name or Material Nickel plating solution, Bright finish

    GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H315-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prol

    GHS P Statement
    P260-P284a-P261-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container to local regulations

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit