Nickel Powder, 75% on graphite, <75 micron, Alfa Aesar™

Form: Apparent density 0.8-1.5g/cm3

Manufacturer: Alfa Aesar 1055522

Quantity 100g

 View more versions of this product

Catalog No. AA1055522

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Powder, 75% On Graphite
    MDL Number MFCD00011137
    Chemical Name or Material Nickel

    Specifications

    Physical Form Powder, 75% On Graphite
    Quantity 100g
    Color Black
    DOT Information DOT Class: 4.2, Packaging Group: II
    MDL Number MFCD00011137
    Odor Odorless
    Health Hazard 1 H228-H251-H351-H319-H317-H335 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation
    Appearance Black powder
    Chemical Name or Material Nickel

    H228-H251-H351-H319-H317-H335 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit